Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Begacestat, Tocris Bioscience™

Catalog Number 428310
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
769169-27-9
C9H8ClF6NO3S2
391.727
PSXOKXJMVRSARX-SCSAIBSYSA-N
begacestat, unii-3666c56bbu, 5-chloro-n-1s-3,3,3-trifluoro-1-hydroxymethyl-2-trifluoromethyl propyl-2-thiophenesulfonamide, begacestat usan:inn, begacestat usan/inn, d09zvc, begacestat hplc, 2s-n-5-chlorothiophen-2-yl-4,4,4-trifluoro-1-hydroxy-3-trifluoromethyl butane-2-sulfonamido, 2-thiophenesulfonamide, 5-chloro-n-1s-3,3,3-trifluoro-1-hydroxymethyl-2-trifluoromethyl propyl, 5-chloro-n-1s-3,3,3-trifluoro-1-hydroxymethyl-2-trifluoromethyl propyl thiophene-2-sulfonamide
11269353
5-chloro-N-[(2S)-4,4,4-trifluoro-1-hydroxy-3-(trifluoromethyl)butan-2-yl]thiophene-2-sulfonamide
C1=C(SC(=C1)Cl)S(=O)(=O)NC(CO)C(C(F)(F)F)C(F)(F)F
This item is not returnable. View return policy
769169-27-9
C9H8ClF6NO3S2
391.727
PSXOKXJMVRSARX-SCSAIBSYSA-N
begacestat, unii-3666c56bbu, 5-chloro-n-1s-3,3,3-trifluoro-1-hydroxymethyl-2-trifluoromethyl propyl-2-thiophenesulfonamide, begacestat usan:inn, begacestat usan/inn, d09zvc, begacestat hplc, 2s-n-5-chlorothiophen-2-yl-4,4,4-trifluoro-1-hydroxy-3-trifluoromethyl butane-2-sulfonamido, 2-thiophenesulfonamide, 5-chloro-n-1s-3,3,3-trifluoro-1-hydroxymethyl-2-trifluoromethyl propyl, 5-chloro-n-1s-3,3,3-trifluoro-1-hydroxymethyl-2-trifluoromethyl propyl thiophene-2-sulfonamide
11269353
5-chloro-N-[(2S)-4,4,4-trifluoro-1-hydroxy-3-(trifluoromethyl)butan-2-yl]thiophene-2-sulfonamide
C1=C(SC(=C1)Cl)S(=O)(=O)NC(CO)C(C(F)(F)F)C(F)(F)F

γ-secretase inhibitor; lowers Aβ40 and Aβ42 levels

γ-secretase inhibitor; selectively inhibits cleavage of amyloid precursor protein (APP) over Notch. Lowers levels of Aβ42 and Aβ40 (EC50 values are 12.4 and 14.8 nM respectively in cells expressing human recombinant APP). Orally active.
Quantity 10 mg
Formula Weight 391.74
Percent Purity >98%
Chemical Name or Material Begacestat
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit