Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ Aero™ Emergency Eye Wash Solution; Buffered pH Neutralizing, Stations and Bottles
Bel-Art™ Aero™ Emergency Eye Wash Solution provides immediate first aid for the eyes. Effective in rinsing impurities and irritants from the eye and in preventing damage from acids and corrosive substances
Supplier: Bel-Art™ F248825016
Alert:
These products are available only in the United States.
Description
Features:
- Immediate response eye first aid for the laboratory, classroom, work site and public facilities
- pH neutralizing solution helps the eye return to a normal 7.4pH level by quickly neutralizing corrosive chemicals
- Instant access to fast, safe, and effective first aid for eye injuries caused by foreign particles such as dirt and debris, chemicals and other environmental irritants
- All-in-one eye wash combines sterile saline with buffered, pH neutralizing solution to help relieve irritation, discomfort and minimize likelihood of permanent damage
- 4 year shelf life from date of manufacture
- Meets all FDA requirements for eye wash including buffered pH solution and preservative as per 21 CFR part 349
- Unique eye-opener allows used fluids and contaminants to flow away from the eye (16oz and 32oz bottles)
- Covered station keeps bottles clean throughout shelf life; carrying handle makes transport easy for field work; wall mountable, hardware included
- Smaller 5oz. and 0.5oz. sizes can be easily included in portable work stations or in pockets for high risk situations
- Quick twist top; easy-to-open and use
- Meets ANSI Z358.1-2009 (as a personal eye wash only)
Specifications
16 oz. | |
Clear | |
Unique Eye-opener |
473 mL | |
Rinsing Impurities and Irritants from the Eye; Preventing Damage from Acids and Corrosive Substances | |
Eyewash Station |
Safety and Handling
ShelfLife : 4 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction