Bel-Art H-B™ DURAC™ Four Channel Timer with White Board
Four channel timer has built in white board that helps keep track of time and other critical information
Manufacturer: H-B Instrument CompanySupplier Diversity Partner B60208-0200
 View all options for this product
Catalog No. 13-202-735
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Includes Timer with White Board; Indiv. calibration report, Traceable to NIST
    Battery Type Two 1.5V, type AAA or equivalent (included)
    Product Type With individual calibration report, Traceable to NIST
    Resolution One second
    Quantity 3/Cs.
    Timer ±Five seconds/day
    • Audible alarm for each channel
    • Built in white board and dry erase marker with eraser tip (included) helps keep track of crucial information
    • Programs: hours/minutes; Displays: hours/minutes/seconds
    • Switchable timing capacity: 23 hours: 59 minutes / 59 minutes
    • Count direction: up/down; displays elapsed time after countdown
    • Last time set memory
    • Key press confirmation feature gives an audible indication of data entry
    • Lay flat on a bench, use the flip-open stand for a vertical view, or use magnetic back for metal surface attachment
  • Description & Specifications
Related Products
Product Content Feedback