Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Bel-Art™ SP Scienceware™ PTFE Disposable Stir Bars PROMO
Octagon magnetic stirring bars with integral pivot ring
Manufacturer:  Bel-Art™ 838874026
 View more versions of this product
Catalog No. 22-067-157PMO
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Yes
PTFE
Alnico Magnet
0.375 in
9.5mm
FDA, USP Class VI, ISO 9001:200
White
Octagon
0.875 in.
22mm