Learn More
Bel-Art™ SP Scienceware™ PTFE Disposable Stir Bars PROMO
Octagon magnetic stirring bars with integral pivot ring
Manufacturer:  Bel-Art™ 838874025
 View more versions of this product
Catalog No. 22-066-910PMO
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Yes
PTFE
Alnico Magnet
0.31 in.
8mm
FDA, USP Class VI, ISO 9001:200
White
Octagon
0.87 in.
22mm