Learn More
Bel-Art™ SP Scienceware™ Lab-Aire™ II Glassware Dryers Accessories Available on GSA/VA Contract for Federal Government customers only.
Replacement pegs for use with Lab-Aire II benchtop and wallmount dryer models.
$46.55 - $49.90
Specifications
For Use With (Equipment) Lab-Aire II benchtop and wallmount dryers
Type Replacement Peg
View More Specs
Products 3
Catalog Number Mfr. No. Length (English) Size Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Size Price Quantity & Availability  
03-422-208
View Documents
Bel-Art™
F189310002
4 in. Medium Pack for $48.40
Add to cart
03-422-207 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Bel-Art™
F189310001
3 in. Small Pack for $46.55
Add to cart
03-422-209 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Bel-Art™
F189310003
5 in. Large Pack for $49.90
Add to cart
Description
Description
    • Available as small (3 in.), medium (4 in.) and large (5 in.) pegs
    • Pegs can be moved and repositioned to hold variety of labware including beakers, flasks, bottles, test tubes and cylinders
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Lab-Aire II benchtop and wallmount dryers
Replacement Peg