Learn More
Bel-Art™ SP Scienceware™ V-Channel Pegs for Lab-Aire™ II Glassware Drying Racks Products for Science Education
Use with Bel-Art™ Lab-Aire™ II labware drying racks. SP Scienceware™ V-Channel Pegs & Plugs are used in place of traditional pegs to increase air flow.
Manufacturer:  Bel-Art™ F189310001/EMD
 View more versions of this product
Catalog No. S93967
Add to cart
Description
Description
  • V-channel pegs/plugs can be repositioned to hold a variety of labware, including beakers, flasks, bottles, test tubes and cylinders
  • Rugged ABS plastic pegs wipe clean and will not corrode
  • Three sizes and lengths: small (3 in.), medium (4 in.) and large (5 in.)
  • Pack includes 19 pegs and 6 plugs
Specifications
Specifications
Not Autoclavable
3 in.
ABS Plastic
Dryers
7.5 cm
Accessory