Learn More
Bel-Art™ SP Scienceware™ Lab-Aire™ II Dryer Accessories Available on GSA/VA Contract for Federal Government customers only.
Replacement pegs for use with Lab-Aire II benchtop and wallmount dryer models.
Manufacturer:  Bel-Art™ F189310002
 View more versions of this product
Catalog No. 03-422-208
  Check Availability
Add to cart
Specifications
Description
  • Available as small (3 in.), medium (4 in.) and large (5 in.) pegs
  • Pegs can be moved and repositioned to hold variety of labware including beakers, flasks, bottles, test tubes and cylinders
Description & Specifications
Specifications
No
4 in.
Replacement peg
Lab-Aire II benchtop and wallmount dryers
Medium