Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ PrepSafe™ Microcentrifuge Tube Mini Floating Racks

Immobilizing tabs protect tubes from opening when under pressure, submerged or knocked over

Manufacturer:  Bel-Art™ F187420140

 View more versions of this product

Catalog No. 03-410-719


  Check Availability
Add to cart

Description

Description

Immobilizing tabs protect tubes from opening when under pressure, submerged or knocked over

  • Holds up to twelve 1.5mL or 2.0mL microcentrifuge tubes in numbered locations
  • Raised handle for easy lifting without agitating samples
  • Stable feet keep racks upright on the bench
  • Use in water baths, ice or on the benchtop
  • 95mm outer diameter fits most 1000mL beakers
  • Available with or without vortexing attachment in a variety of colors
  • Polypropylene construction is steam autoclavable at 121°C (250°F)
  • Removable vortexing attachment available separately (Mfr. No. F187420140)
Specifications

Specifications

Yes
For samples to be vortexed simultaneously
1.5 to 2mL Tubes
1
Vortexing Attachment
Natural
Microcentrifuge tube mini floating racks
Polypropylene
1 Ea.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit