Bel-Art™ SP Scienceware™ Adjustable Elution Rack
Perform as many as 42 elutions in one rack, saving valuable bench space
Manufacturer: Bel-Art™ F188820002
 View more versions of this product
Catalog No. 11-999-008
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Autoclavable Yes
    Type Elution Rack
    Holds 16mm Tubes
    Positions 30.0,30.0
    Material Polypropylene
    Product Type Elution Rack
    Tube Size 16mm
  • Description & Specifications
Related Products
Product Content Feedback