Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ SP Scienceware™ Bacterial Cell Spreaders
Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.
Supplier: Bel-Art™ F377360006
Description
Spreads bacterial cells onto the surface of an agar Bel Art™ SP Scienceware™ Bacterial Cell Spreaders are available in two sizes.
- Autoclavable and may be flame sterilized
- Features triangular end with smooth rounded surfaces throughout
- Smaller 16mm width bar suitable for use with 6cm plates
- Larger 25mm width bar suitable for use with 9cm plates
- Use with Inoculating Turntable (Mfr. No.F40913-0000)
- Both sizes 30cm long
Specifications
30.2 cm | |
Stainless Steel | |
Yes | |
Bacterial Cell Spreader |
Sterile | |
12 in. | |
Inoculating turntable, 6cm inoculating plates | |
Easily perform inoculation of petri dishes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction