Learn More
Bel-Art™ SP Scienceware™ Battery-Powered Engraving Tool and Stand
Permanently mark important tools, samples and more in seconds
Manufacturer:  Bel-Art™ F441500015
 View more versions of this product
Catalog No. 08-647-871
Add to cart
Requires : “AA” batteries (not included)
Description
Description
Engraving Tool (Mfr. No. F441500005)
  • Hold the Engraving Tool like a pen, press the power button, apply light pressure, and “write” to permanently mark most materials including glass, plastic, wood, metal, bone, and stone
  • Portable, cordless battery power: operates on two “AA” batteries (not included)
  • Use indoors or out in the field
  • Marks will not fade if exposed to chemicals or sunlight
Engraving Tool Stand (Mfr. No. F441500015)
  • Keeps tool handy when not in use
  • Molded from polyethylene
  • Well to hold extra engraving tip
Specifications
Specifications
Battery powered engraving tool stand
Extra engraving tip
Polypropylene