Learn More
Bel-Art™ SP Scienceware™ Bellows Type Glove Box Gloves
Forms a leak-proof unit with the box. Made from natural latex with a removable hand, the sleeves fit 20cm (8 in.) circular ports and 18 x 25cm (7 x 10 in.) oval ports.
Supplier:  Bel-Art™ F500250332
 View more versions of this product
Catalog No. 11-391-51
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
This is a replacement glove only. 
  • Forms a leak-proof unit with O-ring construction integral with the box
  • Bellows allows cooling air circulation
  • When the glove and sleeve are separated, the sleeve remains attached to the box
  • Glove sizes are interchangeable, and almost any standard laboratory, surgical or disposable glove can be used
  • Length of the extended sleeve and glove is approximately 76cm (30 in.)
  • Use with H50025-0320 Clamping Rings to hold gloves in place
Note: Due to the unspecified nature of their use and the environment in which they are used, glove box gloves may only be returned due to shipping damage, and only if they have not been used.
Order Info
Sleeve fits 8 in. O.D. (20cm) circular ports and 7 x 10 in. (18 x 25cm) oval ports. Extended length with glove, 30 in. (76cm). One pair per pack.
Specifications
Specifications
Replacement Glove
Natural Rubber, Latex
SP Scienceware™ Budget-Priced Glove Box
Medium (9)
Product Suggestions
Product Suggestions