Bel-Art™ SP Scienceware™ Budget-Priced Glove Box: Gloves with Sleeves
For use with Budget-priced Glove Box
Manufacturer: Bel-Art™ H500250309
 View more versions of this product
Catalog No. 11-391-105B
  Check Availability
Add to Cart
Alert
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
    Specifications
    Material Natural Rubber, Neoprene
    Type Gloves with sleeves
    For Use With (Application) Changing of hands for different sizes or types of hands on same bellows sleeves
    For Use With (Equipment) Budget-priced glove box with 8 in. O.D. circular ports and 7 x 10 in. oval port
    Size 9
  • Description & Specifications
Related Products
Product Content Feedback