Learn More
Bel-Art™ SP Scienceware™ Carboys with Spigot Available on GSA/VA Contract for Federal Government customers only.
Lightweight, durable polyethylene with extra-heavy walls for long life and dependability
$204.50 - $338.50
Specifications
Material Polyethylene
For Use With (Application) For dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes
Type Carboy
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
01-189-0112 View Documents Bel-Art™
F118470010
1 gal. 4 L Each for $204.50
Case of 6 EA for $1,097.00
Add to cart
01-189-0113 Available on GSA/VA Contract for Federal Government customers only. View Documents Bel-Art™
F118470020
2 gal. 8 L Each for $233.50
Case of 6 EA for $1,224.00
Add to cart
22-290047 Available on GSA/VA Contract for Federal Government customers only. View Documents Bel-Art™
F118470050
5 gal. 18.9 L Each for $338.50
Case of 4 EA for $1,252.00
Add to cart
Description
Description
  • Lighter than glass and easy to handle
  • Heavy knurling on cap provides excellent grip for opening and closing
  • Resistant to corrosive acids and alkalis
  • The spigot requires only half turn to open or close
  • Smooth sides accept adhesive labels and can be marked with most indelible markers
Use to dispense cleaning solutions, laboratory reagents, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Polyethylene
For dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes
Carboy