Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Polyethylene, Carboy with Spigot

Click to view available options
Capacity (Metric):
4 L
20 L

For Use With: For dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes; Closure Material: Polyethylene; Color: White

  • Lighter than glass and easy to handle
  • Heavy knurling on cap provides excellent grip for opening and closing
  • Resistant to corrosive acids and alkalis
  • The spigot requires only half turn to open or close
  • Smooth sides accept adhesive labels and can be marked with most indelible markers

Use to dispense cleaning solutions, laboratory reagents, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes.

Color White
Closure Material Polyethylene
Material Polyethylene
Type Carboy with Spigot
Capacity (English) 1 gal.
For Use With (Application) For dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes
Capacity (Metric) 4 L