Learn More
Bel-Art™ Cloning Discs
Save time and reduce effort when isolating a large number of clones
Manufacturer:  Bel-Art™ F378470002
 View more versions of this product
Catalog No. 07-907-10B
Add to cart
Description
Description
Save time and reduce effort when isolating a large number of clones
  • The sterile paper discs transfer cells quickly and easily to a 24-well plate
  • Cloning discs have been sterilized by Gamma radiation to be RNase and DNase free
Order Info
Supplied in vials of 50, two vials per pack.
Specifications
Specifications
RNase and DNase free
4.8mm
Sterile
Cloning Disc
0.18 in.
Ideal for cell transfer and propagation
Paper