Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Disposable Cartridge

Material: Charcoal; For Use With: Incubator

Supplier:  Bel-Art™ F249890000

Catalog No. 03-420-113

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Lessens chance of breathing volatile component
  • Reduces contamination of incubator and water in humidity pan
  • Place cartridges on incubator shelf and, after use, dispose in regular beta waste containers
  • Cartridges are sealed with a highly porous durable membrane

Specifications

Specifications

Incubator
Cartridge
3.9 in.
Charcoal
Incubator
10 cm
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit