Learn More
Bel-Art™ SP Scienceware™ Disposable Pipetting Reservoirs
Disposable and economical plastic pipetting reservoir. The SP Scienceware™ Disposable Pipetting Reservoirs includes ample clearance for all types of pipettors.
Manufacturer:  Bel-Art™ F378770001
 View more versions of this product
Catalog No. 14-512-65
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Ample clearance for all types of pipettors
  • Used for extended multichannel pipetting into microwell plates, Mini-Tubes, Cube 2ube™ Tubes and deep well blocks
  • 100ml capacity
Specifications
Specifications
Sterile
6.12 x 3.62 x 1.25 in. (15.54 x 9.19 x 3.17cm)
1.25 in.
6.12 in.
Individually Wrapped
9.19 cm
Plastic
Pipetting Reservoir
Yes
3.17 cm
15.54 cm
3.62 in.
100 mL