Learn More
Bel-Art™ SP Scienceware™ Disposable Pipetting Reservoirs Available on GSA/VA Contract for Federal Government customers only.
Disposable and economical plastic pipetting reservoir. The SP Scienceware™ Disposable Pipetting Reservoirs includes ample clearance for all types of pipettors.
Manufacturer:  Bel-Art™ F378770000
 View more versions of this product
Catalog No. 13-712-14
  Check Availability
Add to cart
Specifications
Description
  • Ample clearance for all types of pipettors
  • Used for extended multichannel pipetting into microwell plates, Mini-Tubes, Cube 2ube™ Tubes and deep well blocks
  • 100ml capacity
Description & Specifications
Specifications
No
100mL
3.17cm
3.38 oz., 100mL
6.12 in.
15.54cm
9.19cm
3.38 oz.
1.25 in.
Bulk
Yes
Pipetting Reservoir
3.62 in.
6.12 x 3.62 x 1.25 in. (15.54 x 9.19 x 3.17cm)