Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Bel-Art™ SP Scienceware™ DUO Eyewash Station
Bel-Art DUO wall-mountable eyewash stations with sterile saline and bottles of pH neuralizing DUO eye wash are ideal for areas where there is risk of injuries from acids and alkalis is high.
Manufacturer:  Bel-Art™ F248808010
 View more versions of this product
Catalog No. 19-991-073
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
33.81 oz.
33.81 oz., 1000mL
1000mL 0.9% Sterile Saline bottle, placard each with easy to follow universal pictograms, a polished metal mirror and all mounting hardware, built-in eye piece
Open station eyewash bottle
Yes
1000mL
ANSI Z358.1-2009, CE
Wall
5 min./bottle
5 to 35°C
Safety and Handling
ShelfLife : 3 Years