Learn More
Bel-Art™ SP Scienceware™ DUO Eyewash Station
Bel-Art DUO wall-mountable eyewash stations with sterile saline and bottles of pH neuralizing DUO eye wash are ideal for areas where there is risk of injuries from acids and alkalis is high.
Manufacturer:  Bel-Art™ F248808403
 View more versions of this product
Catalog No. 19-991-077
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
16, 33.81 oz.
16, 33.81 oz., 500, 1000mL
1000mL Saline bottle and 500mL pH neutralizing solution, pictograms, placard, hardware, built-in eye piece, polished Metal mirror, mounting hardware
Open station eyewash bottle
Yes
500, 1000mL
ANSI Z358.1-2009, CE
Wall
5 min./500mL Bottle and 2 min./200mL bottle
5 to 35°C
Safety and Handling
ShelfLife : 3 Years