Learn More
Bel-Art™ SP Scienceware™ Earth Friendly 1001 Sticks
Lessen environmental impact with polylactic acid (PLA) resin sampling sticks
Manufacturer:  Bel-Art™ F369310001
 View more versions of this product
Catalog No. 14-512-206
Add to cart
Description
Description
The production of 100% plant-based PLA contributes fewer greenhouse gases and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource.
  • Suitable for mixing samples, handling capsules, scraping containers or positioning solid samples for examination
  • Green alternative to petroleum-based sampling tools helps meet sustainability goals
  • Comparable in strength to polystyrene, polypropylene and other petroleum-based plastic resin sampling tools
  • Disposal options include industrial composting for biodegradability, incineration, recycling and landfill
  • Nonsterile FDA-grade: safe for food, drug and cosmetic contact
  • Use a separate stick for each material sampled to avoid cross-contamination
Specifications
Specifications
500/Cs.
FDA
Suitable for mixing samples, handling capsules, scraping containers or positioning solid samples for examination
0.75 in.
5.75 x 0.75 in. (14.6 x 1.9cm)
14.6cm
Non-sterile
Earth-Friendly™ Sampling Stick
5.75 in.
1.9cm
PLA Resin