Learn More
Bel-Art™ SP Scienceware™ Earth Friendly Long-Handle Sampling Spoons
SCI_ED
Lessen environmental impact with PLA (polylactic acid) resin sampling spoons.
$21.75
Specifications
Certifications/Compliance FDA
Product Type Earth Friendly Sampling Spoon
For Use With (Application) Ideal for acquiring samples from bottles, jars and other confined areas and helping to avoid personal contact with sample materials, used as a green alternative to petroleum-based sampling tools
Handle Type Long
Includes Flat edge rim
View More Specs
Products 1
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Price Quantity & Availability  
S04785A
SCI_ED
View Documents
Bel-Art™
F369421000/EMD
0.5 tsp., 2.46mL
Pack of 10 for $21.75
Case of 10 PK for $217.50
Add to cart
Description
Description
The production of 100% plant-based PLA contributes fewer greenhouse gases and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill.
  • FDA-grade spoons safe for food, drug and cosmetic contact
  • Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
  • Green alternative to petroleum-based sampling tools
  • Long handle, 7 in. (17.8cm), helps prevent personal contact with sampled material when reaching into bottles, jars and other confined areas
  • Rim of each bowl has flat edge to assist in collecting and measuring small amounts with accuracy
  • Use of separate spoon for each material sampled to avoid cross contamination
  • Nonsterile
  • Pack of 10 spoons
Specifications
Specifications
FDA
Ideal for acquiring samples from bottles, jars and other confined areas and helping to avoid personal contact with sample materials, used as a green alternative to petroleum-based sampling tools
Flat edge rim
17.8cm
Non-sterile
Earth Friendly Sampling Spoon
Long
7 in.
PLA Resin