Learn More
Bel-Art™ SP Scienceware™ Earth Friendly Wide Handle Sampling Spoons
Lessen environmental impact with PLA (polylactic acid) resin sampling spoons
Manufacturer:  Bel-Art™ F367401025
 View more versions of this product
Catalog No. 14-512-204
Add to cart
Description
Description
The production of 100% plant-based PLA contributes fewer greenhouse gases and uses less nonrenewable energy from source to resin than its petroleum-based counterparts, and the plants used are an annually renewable resource. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill.
  • FDA grade spoons are safe for food, drug and cosmetic contact
  • Comparable in strength and durability to polystyrene, polypropylene and other plastic resin sampling tools
  • Optimized for retrieving small, specific volume samples of powders or liquids
  • Wide handles allows user to grip firmly and provide just enough stiffness to easily sample packed powders and other firm materials
  • Flat bottom shape allows convenient duo use as weighing boats to save time and assure accuracy
  • Nonsterile
Specifications
Specifications
Flat
FDA
Wide
Non-sterile
20mL
0.67 oz., 20mL
Designed for retrieving small, specific volume samples of powders or liquids, used as a green alternative to petroleum-based sampling tools
PLA Resin
0.67 oz.
Earth Friendly Sampling Spoon