Learn More
Bel-Art™ SP Scienceware™ Electrophoresis Fixing Trays
Used to conveniently fix the gel, drain the fixing solution and then set the filter paper on gel in the tray
Manufacturer:  Bel-Art™ F135540000
 View more versions of this product
Catalog No. 15-243A
Add to cart
Description
Description
  • Extraction of the plates is facilitated by 9.5mm square ridges running lengthwise that hold the plate off the tray
  • Also useful in electrophoresis destaining of large gels
  • Drain stopcock in the side with 1.8 meters (6 ft.) of Vikem™ vinyl tubing, 13mm I.D., permits more complete drainage of fluid into receptacle
  • Provides a slower, more controlled drain rate to avoid disturbing the gel
  • 6.4mm (0.25 in.) thick with top stiffening flange and 5 ribs in the bottom to raise the gel and plates
  • Made of polyethylene
  • Hooks on the rim of the tray hold tubing
Specifications
Specifications
White
7.6 cm
Electrophoresis System
Sturdy top stiffening flange, 5 ribs in the bottom, 9.5mm square ridges, drain stopcock, 1.8m Vikem™ Vinyl tubing, hooks
51 cm
2.51 in.
16.14 in.
2.99 in.
20.07 x 16.14 in. (51 x 41 cm)
Fixing Tray
20.07 in.
Polyethylene
0.64 cm
41 cm