Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells
Feature stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
Specifications
Product Type | Dialysis Cell |
---|---|
Material | Acrylic |
Color | Transparent |
Quantity | Each |
For Use With (Application) | Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
08-666-15
|
Bel-Art™
H402600000 |
1 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Made from clear acrylic
- Filler hole screws on each half cavity are fluoropolymer-coated stainless steel
- Volume shown is for one half cell (double for total cell volume)
- A red dot on one half of the equilibrium cell easily identifies the otherwise identical halves
Specifications
Dialysis Cell | |
Transparent | |
Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket |
Acrylic | |
Each |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title