Learn More
Bel-Art™ SP Scienceware™ Equilibrium Dialysis Cells Available on GSA/VA Contract for Federal Government customers only.
Feature stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
Manufacturer:  Bel-Art™ H402620000
 View more versions of this product
Catalog No. 08-666-17
Add to cart
Description
Description
  • Made from clear acrylic
  • Filler hole screws on each half cavity are fluoropolymer-coated stainless steel
  • Volume shown is for one half cell (double for total cell volume)
  • A red dot on one half of the equilibrium cell easily identifies the otherwise identical halves
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
10mL
Stainless steel screws that tighten down on the dialysis membrane also acting as a gasket
Dialysis Cell
Transparent
Acrylic
Each