Bel-Art™ SP Scienceware™ Ergopet™ Bellows Pipetters Available on GSA/VA Contract for Federal Government customers only.

A rugged bellows provides aspirating force without any need for batteries, power cords or vacuum tubing

Manufacturer: Bel-Art™ F379070011

 View more versions of this product

Catalog No. 03-421-508

  Check Availability
Add to Cart

    Specifications

    Autoclavable No
    For Use With (Application) Pipets up to 20mL,Pipets up to 20mL
    Height (English) 11 in.
    Height (Metric) 280mm
    Material Plastic
    Product Type Bellows Pipetter

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit