Learn More
Bel-Art™ SP Scienceware™ Fast-Release Pipette Pump™ II Pipetters
Includes a rapid release valve for quick dispensing. Bel Art™ SP Scienceware™ Fast-Release Pipette Pump™ II Pipetters are color coded by size for easy identification.
Supplier:  Bel-Art™ F379111002
Catalog No. 13-683-1B
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
View alternate product
Description
Description
  • Rotate the knurled thumb wheel to aspirate liquid to the desired volume
  • Apply light pressure to the fast release lever to rapidly dispense
  • Rotate the thumb wheel for gradual dispensing
  • Glass or plastic pipettes are easily inserted into a unique collar with stabilizing fingers for a secure hold
  • Sizes are color coded; can be disassembled for cleaning
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Accessories