Bel-Art™ SP Scienceware™ Fast-Release Pipette Pump™ II Pipetters Available on GSA/VA Contract for Federal Government customers only.

Includes a rapid release valve for quick dispensing. Bel Art™ SP Scienceware™ Fast-Release Pipette Pump™ II Pipetters are color coded by size for easy identification.

Manufacturer: Bel-Art™ F379111010

 View more versions of this product

Catalog No. 13-683-1C

  Check Availability
Add to Cart

    Specifications

    Autoclavable No
    For Use With (Application) Pipets up to 10mL,Pipets up to 10mL
    For Use With (Equipment) 10mL
    Material Polypropylene/Silicone Rubber
    Color Green
    Product Type Fast Release Pipet Filler

     

    • Rotate the knurled thumb wheel to aspirate liquid to the desired volume
    • Apply light pressure to the fast release lever to rapidly dispense
    • Rotate the thumb wheel for gradual dispensing
    • Glass or plastic pipettes are easily inserted into a unique collar with stabilizing fingers for a secure hold
    • Sizes are color coded; can be disassembled for cleaning

     

  • Description & Specifications
  • Accessories
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit