Learn More
Bel-Art™ SP Scienceware™ Fast-Release Pipette Pump™ II Pipetters Available on GSA/VA Contract for Federal Government customers only.
Includes a rapid release valve for quick dispensing. Bel Art™ SP Scienceware™ Fast-Release Pipette Pump™ II Pipetters are color coded by size for easy identification.
Manufacturer:  Bel-Art™ F379111025
 View more versions of this product
Catalog No. 13-683-1D
  Check Availability
Add to cart
Specifications
Description
  • Rotate the knurled thumb wheel to aspirate liquid to the desired volume
  • Apply light pressure to the fast release lever to rapidly dispense
  • Rotate the thumb wheel for gradual dispensing
  • Glass or plastic pipettes are easily inserted into a unique collar with stabilizing fingers for a secure hold
  • Sizes are color coded; can be disassembled for cleaning
Description & Specifications
Specifications
No
25mL
Red
Pipets up to 25mL
Polypropylene/Silicone Rubber
Fast Release Pipet Filler
Accessories