Bel-Art™ SP Scienceware™ Flea Micro Spinbar™ Magnetic Stirring Bars

Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated

Manufacturer: Bel-Art™ F371190635

 View more versions of this product

Catalog No. 22-261681

  Check Availability
Add to Cart

    Specifications

    Shape Octagon
    Diameter (English) 0.118 in.
    Length (English) 0.25 in.
    Diameter (Metric) 3mm
    Length (Metric) 6.35mm
    Autoclavable Yes
    Case Quantity 50
    Certifications/Compliance FDA, USP Class VI, ISO 9001:2008
    Coating PTFE
    Color White
    Material Alnico Magnet
    Product Type Stir Bar

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit