Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Flea Micro Spinbar™ Magnetic Stirring Bars Available on GSA/VA Contract for Federal Government customers only.

Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated

Manufacturer:  Bel-Art™ F371190007

 View more versions of this product

Catalog No. 14-512-36



Add to cart

Description

Description

Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated

  • Small size bar for stirring in vessels such as vials, micro test tubes and centrifuge tubes
  • Alnico magnet encapsulated in FDA Grade
  • USP Class VI PTFE low friction coating
  • Durable and inert
  • Autoclave safe

Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Certifications

Manufactured in a registered ISO 9001:2008, as verified by SGS certification

Specifications

Specifications

Yes
PTFE
0.07 in.
50
2mm
Alnico Magnet
FDA, USP Class VI, ISO 9001:2008
White
0.27 in.
Stir bar
7mm
Octagonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit