Learn More
Bel-Art™ SP Scienceware™ Inoculating Turntable
Allows even distribution of bacterial cells in Petri dishes from 60 through 100mm dia.
Manufacturer:  Bel-Art™ F409130000
Catalog No. 14-512-64
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Lightweight and portable
  • Hand-operated turntable . 
  • Ball bearings ensure smooth and effortless spinning for faster and more accurate inoculation
  • Recessed, non-skid, neoprene turntable top keeps Petri dishes in place
  • Removable top for cleaning
  • Rugged body made of easy-to-clean polypropylene
Specifications
Specifications
Inoculating Turntable
5 in.
60 to 90mm petri dishes
Polypropylene
12.7 x 5cm (5 x 2 in.)
12.7cm
Non-skid neoprene recessed stabilizing pad