Bel-Art™ SP Scienceware™ Klett™ Clinical Colorimeter
Measures in Klett units, directly proportional to the solution's OD, according to Beer's Law
Manufacturer: Bel-Art™ T370120000
 View more versions of this product
Catalog No. 22-065011
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Clinical Colorimeter
    Accuracy 0.33%
    Voltage 115V
    Certifications/Compliance CE
    For Use With (Application) Colorimetric/Turbidimetric analyses in Klett units
  • Description & Specifications
Related Products
Product Content Feedback