Learn More
Bel-Art™ SP Scienceware™ Klett™ Colorimeter Filters
Bel-Art™ SP Scienceware™ Klett™ Colorimeter Filters are glass filters for colorimetric and turbidimetric analyses to obtain readings on the Klett™ scale.
Manufacturer:  Bel-Art™ T370140066
 View more versions of this product
Catalog No. 22-065383
  Check Availability
Add to cart
Specifications
Description
  • Additional Color filters for measurement of entire visual range
  • Klett Colorimeter Clinical and Industrial models are supplied with two most commonly used filters, blue for 400 to 450nm and green for 520 to 580nm
Description & Specifications
Specifications
Klett™ clinical colorimeter, Model 800-3 or Klett™ industrial colorimeter Model 900-16
50 to 70nm
Colorimetric and turbidimetric analysis
51mm
Colorimeter filter
Glass
Red
KS-66
640 to 700nm