Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ SP Scienceware™ Lab Fridge™ Tray Racks and Cabinet
Space-saving storage trays and cabinet eliminate clutter and facilitate stock rotation
Supplier: Bel-Art™ H186610000
Description
- These tray racks eliminate clutter and allow easy stock rotation
- Vacuum-formed white styrene
- Cabinet and Racks sold separately
Lab Fridge Tray Racks
- Suitable for storage of small bottles of blood-typing sera, chemistry controls, enzyme reagents, coagulant reagents, etc.
- Vacuum-formed white styrene is lightweight and easy to clean
- Select 9-channel (Mfr. No. 18661-0000) or 8-channel (18662-0000) tray racks
- 9-channel tray measures 13 x 13 x 1.75 in., with channel width of 1 in.
- 8-channel tray measures 13 x 13 x 1.75 in., with channel width if 1.25 in.
- Open front cabinet holds up to three Lab Fridge Tray Racks; maximum container height 3 3/4 in. (8.3cm)
- Tray racks slide on side rails and can be easily removed for full access
- Cabinets made of rugged polypropylene that is easy to clean and chemical resistant
Specifications
Tray Rack | |
4.4 cm | |
33.02 cm | |
13 in. | |
White | |
13 x 13 x 1.75 in. (33 x 33 x 4.4 cm) | |
1.75 in. |
33 cm | |
13 in. | |
9 | |
Lab Fridge™ tray cabinet | |
Not Autoclavable | |
Space saving storage trays and cabinet eliminate clutter and facilitate stock rotation | |
Polypropylene, Polystyrene |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction