Bel-Art™ SP Scienceware™ Labels for Protective Labeling Tape System

Labels for use with Bel Art Scienceware Protective Labeling Tape System (cat. no. 1186750)

Manufacturer: Bel-Art™ F134550010

 View more versions of this product

Catalog No. 11-867-19

  Check Availability
Add to Cart

    Specifications

    Format Label
    For Use With (Application) Use to label labware, bottles or samples
    Length (English) 3 in.
    Length (Metric) 7.62cm
    Material Paper
    Product Type Label
    Quantity 1000/Pk.
    Color White
    Width (English) 1 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit