Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Magic Touch 2™ Ice Buckets Available on GSA/VA Contract for Federal Government customers only.

Superior insulating properties combined with high chemical resistance

Manufacturer:  Bel-Art™ M168072001

 View more versions of this product

Catalog No. 11-999-054


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

0.56 gal., 2.5L
10.56 in.
Lightly textured, nonslip
-320° to 212°F
Raised
Ice Bucket
2.5L
17.9cm
Blue
26.8cm
Pour spout, drip-proof rims, fill line indicator, snug-fitting keyed lid and base
-196° to 100°C
Urethane
0.56 gal.
7.06 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit