Learn More
Bel-Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans Available on GSA/VA Contract for Federal Government customers only.
Keep temperature sensitive samples in an optimal environment. Bel Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans are the lightest icewares on the market.
Manufacturer:  Bel-Art™ M188489101
 View more versions of this product
Catalog No. 11-675-123
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Blue
5.1 in.
16 in.
13 in.
9L
Ideal for storing and transporting temperature sensitive samples in Ice, Alcohol Slurries, Water baths and more
-320.8 to 199.4°F
13cm
41cm
33cm
16 x 13 x 5.1 in. (41 x 33 x 13cm)
Maxi Lab Ice Pan
Polymer Foam
-196 to 93°C