Learn More
Bel-Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans
Keep temperature sensitive samples in an optimal environment. Bel Art™ SP Scienceware™ Magic Touch™ Insulated Lab Pans are the lightest icewares on the market.
Manufacturer:  Bel-Art™ M188481104
 View more versions of this product
Catalog No. 11-675-131
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Green
3 in.
6 in.
6 in.
1L
Ideal for storing and transporting temperature sensitive samples in Ice, Alcohol Slurries, Water baths and more
-320.8 to 199.4°F
7.6cm
15.2cm
15.2cm
6 x 6 x 3 in. (15.2 x 15.2 x 7.6cm)
Mini Lab Ice Pan
Polymer Foam
-196 to 93°C