Learn More
Bel-Art SP Scienceware Mercury Collector
Safely clean up and dispose of dangerous mercury spills.
$40.75 - $41.75
Catalog Number | Mfr. No. | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
S40734
![]() |
Bel-Art
F362500000/EMD |
Mercury Collector | For mercury spill clean up |
Each for $41.75
|
|
S40734A
![]() |
Bel-Art
F362510000/EMD |
Replacement Pad | For mercury collector |
Pack of 12 for $40.75
|
|
Description
- Polyethylene device makes it easy to collect mercury
- Press the lid firmly against surface to pick up individual mercury globules
- Screw lid onto container to wring out mercury; repeat as needed to collect all spilled mercury
- Tight-fitting lid eliminates harmful mercury vapors
- Mercury is collected at the bottom of the container
Specifications
1.88 in. | |
4.77cm |