Learn More
Bel-Art™ SP Scienceware™ Micro-Mill™ Grinder Blades
Replacement blades for MicroMill grinder
Manufacturer:  Bel-Art™ H372570000
 View more versions of this product
Catalog No. 08-335-2
Add to cart
Description
Description
Stainless-steel or hard-faced blades.
Certifications
CE Marked
Specifications
Specifications
Ideal for processing minerals, pharmaceuticals, pigment blends and other solid samples
12000rpm
Replacement Blade
Stainless Steel
MicroMill grinder