Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Microcentrifuge Tube Sample Pestles

Designed to homogenize samples in a microcentrifuge tube

Specifications

Autoclavable Yes
For Use With (Application) Designed to homogenize samples in a microcentrifuge tube
Includes Teflon™ PTFE pestle, Stainless Steel shaft
Length (English) 5.62 in.
Length (Metric) 143mm
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
05-559-26 missing translation for 'viewDocuments' Bel-Art™
F199220000
1.5mL Microcentrifuge tube with standard cone This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
05-559-27 missing translation for 'viewDocuments' Bel-Art™
F199220001
1.5mL Microcentrifuge tube with narrow cone This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Suitable for resuspending lyophilized products or precipitates
  • fluoropolymer pestle is mounted on a stainless steel shaft, 3.2mm (0.125 in.) in diameter
  • May be hand-held or mounted in a motorized chuck
  • Overall length 14.3cm (5.625 in.)
  • Steam autoclavable at 121°C (250°F)
  • Choose rounded or flat tip shape to match centrifuge type

Specifications

Specifications

Yes
Teflon™ PTFE pestle, Stainless Steel shaft
143mm
Designed to homogenize samples in a microcentrifuge tube
5.62 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit