Bel-Art™ SP Scienceware™ Microcentrifuge Tube Sample Pestles Available on GSA/VA Contract for Federal Government customers only.

Designed to homogenize samples in a microcentrifuge tube

Manufacturer: Bel-Art™ F199220001

 View more versions of this product

Catalog No. 05-559-27

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) 1.5mL Microcentrifuge tube
    Includes Teflon™ PTFE pestle, Stainless Steel shaft
    Length (English) 5.62 in.
    Length (Metric) 143mm
    Autoclavable Yes
    For Use With (Application) Designed to homogenize samples in a microcentrifuge tube

  • Description & Specifications
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit