Learn More
Bel-Art™ SP Scienceware™ Microtube Cutter
Safely, accurately and easily sections microcentrifuge tubes and eliminates the hazards of using razor blades or knives
$23.35 - $58.20
Specifications
For Use With Microtube cutter
View More Specs
Products 2
Catalog Number Mfr. No. Material Type Price Quantity & Availability  
Catalog Number Mfr. No. Material Type Price Quantity & Availability  
05-401A
View Documents
Bel-Art™
F210180000
Chrome-plated Steel Microtube Cutter Each for $58.20
Add to cart
05-401B
View Documents
Bel-Art™
F210180001
Steel Microtube Cutter Replacement Blade Each for $23.35
Add to cart
Description
Description
  • Makes it easy to retrieve radioactive pellets from the tips of tubes without resuspension or the risk of splashed radioactive substances
  • Works well with polyethylene c or polypropylene microcentrifuge tubes up to 1.5mL
  • Chrome-plated body with replaceable steel blade
  • Maximum cut diameter is 9.4mm (0.370 in.)
Not recommended for polystyrene tubes.
Specifications
Specifications
Microtube cutter