Learn More
Bel-Art™ Octagon Magnetic Stirring Bar Assortments
Popular sizes and shapes
Manufacturer:  Bel-Art™ F371820000
 View more versions of this product
Catalog No. 22-331462
Add to cart
Description
Description
Popular sizes and shapes
Specifications
Specifications
FDA
White
6
Alnico Magnet
PTFE
12 standard bars two each of 0.5 x 0.31 in., 0.62 x 0.31 in., 1 x 0.37 in., 1.5 x 0.37 in., 2 x 0.37 in., 2.5 x 0.37 in.
Stir bar
Octagonal