Learn More
Bel-Art™ SP Scienceware™ OdoClave™ Deodorant Autoclave Pads Available on GSA/VA Contract for Federal Government customers only.
Neutralize autoclave odors with a pleasant fragrance
Manufacturer:  Bel-Art™ H131980000
 View more versions of this product
Catalog No. 14-488-30
Add to cart
Description
Description
  • Heat-release deodorant works automatically at autoclave temperatures
  • Open pouch and place in bag prior to autoclaving
  • Three-year shelf life or longer if stored in a sealed container
  • 32mm (1.75 in.) square
  • Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Choose 1.25 in. (32mm) square regular-strength or 2.25 in. (5.7cm) square super-strength pad.
Specifications
Specifications
Clove
Yes
Brown