Learn More
Bel-Art™ SP Scienceware™ Pipette Pump™ Pipetters Available on GSA/VA Contract for Federal Government customers only.
Provides safe, accurate and trouble-free pipetting. Bel Art™ SP Scienceware™ Pipette Pump™ Pipetters utilize an easy-to-use one-handed operation.
Manufacturer:  Bel-Art™ F378960000
 View more versions of this product
Catalog No. 13-683A
  Check Availability
Add to cart
Specifications
Description
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
  • Disassemble all parts for cleaning and before autoclaving at 121ºC (250ºF) for 20 minutes; chuck should not be autoclaved
  • Replacement Silicone Rubber Chuck and Washers are available 
Description & Specifications
Specifications
Pipets
Yellow
0.2mL
Acid and Alkali
Pipetter
Accessories